Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
60.10 EUR - 102.66 EUR
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10171901
|
Lenz Laborglasintrumente
05210704 |
750 mm |
102.66 EUR
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
10326171
|
Lenz Laborglasintrumente
05210402 |
400 mm |
60.10 EUR
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
10428792
|
Lenz Laborglasintrumente
05210504 |
500 mm |
65.03 EUR
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
Description
- Liebig condenser acc. to DIN 12576 and in addition to DIN
- For condensing vapours
- With NS socket and NS cone
- Water connection: glass olives
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) |