Learn More
Lenz Laborglasinstrumente™ Condenser
missing translation for 'orderingAttributeHoverText'
Standard Taper Joint No:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
Length (Metric):
160 mm
250 mm
400 mm
missing translation for 'unitSize'
Each
Description
- Chemical resistance
- High temperature resistance
Specifications
Specifications
| Length (Metric) | 250 mm |
| Standard Taper Joint No | NS 24/29 |
| Type | Condenser |