Learn More
Motic™ Condenser for AE2000
Click to view available options
For Use With (Equipment):
AE2000 Microscopes
Unit Size:
Each
Description
- Concentrates and focuses light passing through the specimen.
- Short working-distance condenser (28mm).
Specifications
Specifications
| Description | NA: 0.50 |
| Product Type | Condenser |
| For Use With (Equipment) | AE2000 Microscopes |
| Working Distance | 28 mm |