Learn More
Motic™ Condenser, Achromatic, LWD, BA410
Product Code. 15425302
Change view
Click to view available options
For Use With (Equipment):
BA410 Series Upright Compound Microscope
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15425302 BA410 Series Upright Compound Microscope Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15425302 Supplier Motic™ Supplier No. 1101001901261
Long working distance achromatic condenser suitable for phase contrast. Motic™ Condenser, Achromatic, LWD, BA410 provides spherical and chromatic aberration correction, and facilitates visualization of transparent specimens when used with phase accessories.
  • Concentrates and focuses light passing through the specimen.
  • Long working-distance condenser.
  • Suitable for phase contrast.
  • Spherical and chromatic aberration correction.
Specifications
Type Condenser
Description NA: 0.65, WD: 10.8mm
For Use With (Equipment) BA410 Series Upright Compound Microscope