Learn More
Motic™ Condenser, Achromatic, LWD, BA410
Description
- Concentrates and focuses light passing through the specimen.
- Long working-distance condenser.
- Suitable for phase contrast. Spherical and chromatic aberration correction.
Specifications
Specifications
| Type | Condenser |
| Description | NA: 0.65, WD: 10.8mm |
| For Use With (Equipment) | BA410 Series Upright Compound Microscope |