Learn More
3M™ Comply™ Lead-Free Indicator Tape
3M™ Comply™ Lead-Free Indicator Tape securely seals bags and provides visual proof that the bag has undergone the steam sterilization process.
Brand:  3M™ 7000138231
Product Code. 15804547
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Securely seal bags and allow CSA and OR staff to quickly check whether the bags have undergone the sterilization process
  • The chemical indicator ink, applied in a diagonal striped pattern, changes color or becomes darker when exposed to the sterilization process
  • Due to the use of unleaded ink there are no problems with hazardous waste
Specifications
Specifications
Exposure monitoring
Indicator Tape
48 mm
55 m
Steam
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions