Learn More
3M™ Comply™ Lead-Free Indicator Tape
3M™ Comply™ Lead-Free Indicator Tape securely seals bags and provides visual proof that the bag has undergone the steam sterilization process.
Brand: 3M™ 7000138231
This item is not returnable.
View return policy
Description
- Securely seal bags and allow CSA and OR staff to quickly check whether the bags have undergone the sterilization process
- The chemical indicator ink, applied in a diagonal striped pattern, changes color or becomes darker when exposed to the sterilization process
- Due to the use of unleaded ink there are no problems with hazardous waste
Specifications
| Exposure monitoring | |
| Indicator Tape | |
| 48 mm |
| 55 m | |
| Steam |