missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Complete Dispense System
Shop All Agilent BioTek Products

Click to view available options
Unit Size:
Each
Capacity (Metric):
10 L
Description
- Contains bottles, pumps, and tubing
Specifications
Specifications
| Type | Dispense System |
| For Use With (Equipment) | 406 FX Washer Dispenser |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction