missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C4b/d Antibody (16D2), Novus Biologicals™
Rat Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB200-541
This item is not returnable.
View return policy
Description
Complement C4b/d Monoclonal specifically detects Complement C4b/d in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| Complement C4b/d | |
| Monoclonal | |
| 0.1 mg/mL | |
| Western Blot 1:10, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10, Immunoassay | |
| basic C4, Basic complement C4, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3, C4B1, C4B12, C4B2, C4B3, C4FMGC164979, CH, Chido form of C4, CO4C4B5, complement C4-B, complement C4B1a, complement component 4B, complement component 4B (Chido blood group), CPAMD3FLJ60561, EC 2.1.1.144, EC 2.7.11 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 721 | |
| Mouse | |
| Ascites |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoassay | |
| 16D2 | |
| Unconjugated | |
| P01029 | |
| C4B | |
| Cell-bound C4 of mouse origin. | |
| 0.05 mg | |
| Primary | |
| Antibody cross reacts with C4, C4b and C4d. | |
| Store at 4°C. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction