missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3a Antibody (K13/16-5.7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
222.00 EUR - 531.00 EUR
Specifications
| Antigen | Complement C3a |
|---|---|
| Clone | K13/16-5.7 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:10000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:150, Sandwich ELISA Capture |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18728473
|
Novus Biologicals
NBP1-05122 |
100 μg |
531.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18262703
|
Novus Biologicals
NBP1-05122-0.025MG |
0.025 mg |
222.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement C3a Monoclonal specifically detects Complement C3a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA Capture.Specifications
| Complement C3a | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| 718 | |
| Human C3a | |
| Primary | |
| Store at 4C. Do not freeze. |
| K13/16-5.7 | |
| Western Blot 1:100-1:2000, ELISA 1:10000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:150, Sandwich ELISA Capture | |
| Monoclonal | |
| Purified | |
| RUO | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| C3 | |
| IgG1 κ | |
| Protein A or G purified | |
| Specific for human C3, C3a and C3a (desArg) and does not cross-react with C4a or C5a. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title