missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement C3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110861
This item is not returnable.
View return policy
Description
Immunogen sequence: KVRVELLHNP AFCSLATTKR RHQQTVTIPP KSSLSVPYVI VPLKTGLQEV EVKAAVYHHF ISDGVRKSLK VVPEGIRMNK TVAVRTLDPE RLGREGVQKE DIPPADLSDQ VPDTESETRI LLQGTPVAQM TEDAVDAERL KHL Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 83%.
C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.
Specifications
| Complement C3 | |
| Polyclonal | |
| Unconjugated | |
| C3 | |
| Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3d; complement C3; Complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3; complement component C3 alpha-chain; complement component C3a; complement component C3b; complement component C3d; CPAMD1; ENCF-1; ENCF-2; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; I79_008227; LOC100060539; Neutrophil chemotactic factor-1; Neutrophil chemotactic factor-2; Plp; prepro-C3; Unknown (protein for MGC:134562) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 718 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.30 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P01024 | |
| C3 | |
| Recombinant Protein corresonding to Human Complement C3. Recombinant protein control fragment (Product #RP-95392). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction