Learn More
Kern Sohn Compact Laboratory Balance PFB
Click to view available options
Capacity:
120 g
200 g
300 g
600 g
1200 g
2000 g
3000 g
6000 g
Readability:
0.001 g
0.01 g
0.05 g
0.1 g
Unit Size:
Each
Description
- Easy to use: All primary functions have their own key on the keypad
- Compact size, practical for small spaces
- Capacity display: A bar graph display lights up to show how much of the weighing range is still available
- Level indicator and levelling feet for precise levelling of the scale, fitted as standard, to give the most accurate weighing result
Specifications
Specifications
| Capacity | 1200 g |
| Readability | 0.01 g |
| Linearity | 0.03 g |
| Type | Compact Precision Balance |
| Electrical Requirements | Universal Mains Adapter |