Learn More
Socorex™ Compact Dispenser 501 Model
Click to view available options
Volume (Metric):
0.2 to 2 mL
0.4 to 5 mL
1 to 10 mL
1 to 30 mL
Increments:
0.1 mL
0.2 mL
1.0 mL
Volume (Metric) Reservoir:
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Specifications
Specifications
| Product Type | Compact Dispenser |
| Autoclavable | Autoclavable |
| Volume (Metric) | 1 to 10 mL |
| Increments | 0.2 mL |
| Model | 501 |
| Volume (Metric) Reservoir | 2000 mL |
| Warranty | 2 Years |