missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Common Elements Mix 2 Standard: Ag, Al, B, Ca, Co, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn @ 100 μg/mL in 5% HNO3, tr. HF, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Common Elements Mix 2 Standard: Ag, Al, B, Ca, Co, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn @ 100 μg/mL in 5% HNO3, tr. HF
Specifications
Specifications
| Chemical Name or Material | Aluminium/Boron/Calcium/Chromium/Cobalt/Copper/Iron/Lead/Magnesium/Manganese/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-42-8, 7440-70-2, 7440-47-3, 7440-48-4, 7440-50-8 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Aluminium/Boron/Calcium/Chromium/Cobalt/Copper/Iron/Lead/Magnesium/Manganese/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 100 μg/mL/Boron 100 μg/mL/Calcium 100 μg/mL/Chromium 100 μg/mL/Cobalt 100 μg/mL/Copper 100 μg/mL/Iron 100 μg/mL/Lead 100 μg/mL/Magnesium 100 μg/mL/Manganese 100 μg/mL/Nickel 100 μg/mL/Phosphorus 100 μg/mL/Potassium 100 μg/mL/Silicon 100 μg/mL/Silver 100 μg/mL/Sodium 100 μg/mL/Tin 100 μg/mL/Titanium 100 μg/mL/Vanadium 100 μg/mL/Zinc 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction