Learn More
R&D Systems™ CometAssay Alkaline Control Cells
Artikelnummer. 30001324
Change view
Click to view available options
Quantity:
10 Tests
Packungsgröße:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30001324 10 Tests Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30001324 Lieferant R&D Systems™ Lieferanten-Nr. 4256010CC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
A set of suspension cell preparations containing different levels of DNA damage to be used as controls with CometAssay™ Kits
  • Stored using a Liquid Nitrogen Storage System
  • Includes:
    • 500 μL of Healthy Control Cells
    • 500 μL of Low Damage Control Cells
    • 500 μL of Intermediate Damage Control Cells
    • 500 μL of High Damage Control Cells
  • These cryopreserved controls are designed to act as controls to standardize and compare alkaline electrophoresis methods between individual users and laboratories
TRUSTED_SUSTAINABILITY
Spezifikation
Quantity 10 Tests
Synonym Apoptosis kinetics, apoptosis kit, apoptosis kit annexin v, apoptosis kit microscopy
Content And Storage Store in liquid nitrogen. Do not use past expiration date.
For Use With (Application) Controls with CometAssay™ Kit