Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1mm Thick
Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
16
20
5
8
9
Volume (Metric) Well:
20 μL
25 μL
35 μL
40 μL
50 μL
60 μL
100 μL
650 μL
Unit Size:
Each
Specifications
Specifications
| Color | White |
| For Use With (Application) | Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations |
| For Use With (Equipment) | OmniPAGE Mini Vertical Gel Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 9 |
| Quantity | 1 Each |
| Thickness (Metric) | 1 mm |
| Volume (Metric) Well | 50 μL |