Learn More
Fisherbrand™ Combs for MultiSUB Maxi Gel Electrophoresis Unit, 1mm Thick
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
16
2 Prep, 2 Marker
20
25
30
36
40
50
Volume (Metric) Sample:
10 μL
11 μL
14 μL
17 μL
21 μL
27 μL
41 μL
72 μL
315 μL
675 μL
Unit Size:
Each
Description
- Recommended for high resolution gels and publication quality data
- Supplied as standard
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | MultiSUB Maxi-Gel Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 2 Prep, 2 Marker |
| Quantity | 1 Each |
| Thickness (Metric) | 1 mm |
| Volume (Metric) Sample | 315 μL |