Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 0.75mm Thick
Product Code. 11883303 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
16
2 Prep, 2 Marker
20
25
4 Prep, 2 Marker
8
Volume (Metric) Sample:
7 μL
10 μL
12 μL
17 μL
20 μL
30 μL
57 μL
118 μL
270 μL
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Sample unitSize
11883303 1 Prep, 1 Marker 270 μL Each
11893303 10 20 μL Each
11863313 12 17 μL Each
11803323 16 12 μL Each
11843323 2 Prep, 2 Marker 57 μL Each
11853323 20 10 μL Each
11823333 25 7 μL Each
11863333 4 Prep, 2 Marker 118 μL Each
11803343 8 30 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11883303
Only null left
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • For tightly resolved bands
  • Recommended for high resolution gels and publication quality data
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Color Black
For Use With (Equipment) Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit
Type Comb
No. of Wells 1 Prep, 1 Marker
Quantity 1 Each
Thickness (Metric) 0.75 mm
Volume (Metric) Sample 270 μL