Learn More
Hanna Instruments™ Combination pH Buffer Solution Kit
Click to view available options
Includes:
500 mL of: pH 4.01 (HI-7004L), pH 7.01 (HI-7007L), pH 10.01 (HI-7010L), Electrode storage solution (HI-70300L), Electrode cleaning solution (HI-7061L)
Unit Size:
Each
Specifications
Specifications
| Includes | 500 mL of: pH 4.01 (HI-7004L), pH 7.01 (HI-7007L), pH 10.01 (HI-7010L), Electrode storage solution (HI-70300L), Electrode cleaning solution (HI-7061L) |
| Quantity | 5 x 500 mL |