Learn More
Hanna Instruments™ Combination pH Buffer Solution Kit
Product Code. 12931850
Click to view available options
Includes:
500 mL of: pH 4.01 (HI-7004L), pH 7.01 (HI-7007L), pH 10.01 (HI-7010L), Electrode storage solution (HI-70300L), Electrode cleaning solution (HI-7061L)
Unit Size:
Each
Product Code. 12931850
Brand: Hanna Instruments™ HI5471011
Only null left
Specifications
Includes 500 mL of: pH 4.01 (HI-7004L), pH 7.01 (HI-7007L), pH 10.01 (HI-7010L), Electrode storage solution (HI-70300L), Electrode cleaning solution (HI-7061L)
Quantity 5 x 500 mL