Learn More
Sartorius Combijet Connector
Shop All Sartorius Products
Click to view available options
:
Combijet Connector
Description
- The Combisart™ System enables you to select the optimal hardware and consumables for your needs in microbiological analysis or particle count in quality assurance.
- Combisart™ features a modular design and field-proven standard accessories to make your choice easier.
- At the heart of the Combisart™ system is a stainless steel manifold designed to accommodate all types of filter holders and funnels such as:
- Ready-to-use units Biosart™ 100 Monitors
- Ready-to-use units Biosart™ 250 Funnels
- Flammable units such as stainless steel funnels for colony counting
- Autoclavable glass filter holders
- Autoclavable polycarbonate
- Sterile venting of each membrane after filtration
- Sterilization according to ISO 8199
- Special polished stainless steel surfaces allow easy cleaning and rinsing
- Filtration of three or six samples in parallel
- Easy pouring out of non-filterable samples
- Equally easy for right- and left-handed users
- Maximum flexibility due to different set-ups
- Space-saving in the autoclave
- Stainless steel 304 - long lifecycle
Specifications
Specifications
| For Use With (Equipment) | Combisart System |
| Type | Connector |