Learn More
Sartorius Combijet Connector
Product Code. 17584326 Shop All Sartorius Products
Click to view available options
:
Combijet Connector
Product Code. 17584326
Brand: Sartorius 1EAS0022
Only null left
Add to basket
Add to basket
The Combijet Connector is a part for the Combisart™ System for microbiological analysis or particle count in quality assurance. The system features a modular design and field-proven standard accessories to make your choice easier.
  • The Combisart™ System enables you to select the optimal hardware and consumables for your needs in microbiological analysis or particle count in quality assurance.
  • Combisart™ features a modular design and field-proven standard accessories to make your choice easier.
  • At the heart of the Combisart™ system is a stainless steel manifold designed to accommodate all types of filter holders and funnels such as:
    • Ready-to-use units Biosart™ 100 Monitors
    • Ready-to-use units Biosart™ 250 Funnels
    • Flammable units such as stainless steel funnels for colony counting
    • Autoclavable glass filter holders
    • Autoclavable polycarbonate
  • Sterile venting of each membrane after filtration
  • Sterilization according to ISO 8199
  • Special polished stainless steel surfaces allow easy cleaning and rinsing
  • Filtration of three or six samples in parallel
  • Easy pouring out of non-filterable samples
  • Equally easy for right- and left-handed users
  • Maximum flexibility due to different set-ups
  • Space-saving in the autoclave
  • Stainless steel 304 - long lifecycle
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Combisart System
Type Connector