Learn More
Cytiva Adapter for XK Columns
Movable adapter gives a uniform flow that maintains the integrity of the packed bed during HPLC operations. Cytiva Adapter for XK Columns is an assembled unit consisting of a top-end cap, a plunger and an adjusting knob. Available for 16, 26 and 50mm diameter XK columns.
325.08 EUR - 480.00 EUR
Specifications
For Use With (Application) | Liquid Chromatography at Low to Medium Pressure |
---|---|
Includes | Plunger, Top-end cap, Adjusting knob |
Quantity | 1 Pack |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11320352
|
Cytiva
28-9898-76 |
XK Empty Column, 16 mm Diameter |
325.08 EUR
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11330352
|
Cytiva
28-9898-77 |
XK Empty Column, 26 mm Diameter |
403.00 EUR
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11340352
|
Cytiva
28-9898-80 |
XK Empty Column, 50 mm Diameter |
480.00 EUR
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
XK empty columns are designed for liquid chromatography at low to medium pressure. They are user friendly, robust, and give reproducible packing, and they feature fittings for direct connection to ÄKTA™ design instruments or other HPLC systems. XK columns are supplied with a movable adapter.
- Plunger design ensures a homogeneous buffer distribution and reproducible packing
- QuickLock mechanism of the adapter shaft facilitates easy column handling and cleaning
- Adapter top-end cap fits directly into the column end piece
- Plunger carries the net ring and support screen for the medium bed
- An adjusting knob seals the adapter to the column wall
Specifications
Liquid Chromatography at Low to Medium Pressure | |
Plunger, Top-end cap, Adjusting knob | |
1 Pack |