missing translation for 'onlineSavingsMsg'
Learn More

Columbin, MedChemExpress

Product Code. 16464827
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464827 50 mg 50mg
16448968 10 mg 10mg
16431398 5 mg 5mg
16423866 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464827 Supplier Medchem Express Supplier No. HYN0389

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Columbin is an orally active diterpenoid furanolactone from Calumbae radix, has anti-inflammatory and anti-trypanosomal effects. Columbin selectively inhibits COX-2 (EC50=53.1 μM) over COX-1 (EC50=327 μM).

Columbin is an orally active diterpenoid furanolactone from Calumbae radix, has anti-inflammatory and anti-trypanosomal effects. Columbin selectively inhibits COX-2 (EC50=53.1 μM) over COX-1 (EC50=327 μM).

Specifications

Chemical Name or Material Columbin
Physical Form Solid
CAS 546-97-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H22O6
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (279.03 mM)
SMILES O=C1[C@](C=C2)(O)[C@@](CC3)(C)[C@@]([C@@](C[C@@H](C4=COC=C4)O5)(C)[C@]3([H])C5=O)([H])[C@]2([H])O1
Molecular Weight (g/mol) 358.39
Formula Weight 358.39
Percent Purity 99.77%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.