Learn More
Thermo Scientific™ Columbia CNA Aesculin Selective Agar / Brilliance™ UTI
Description
Accurately isolate and identify common microorganisms associated with Urinary Tract Infections (UTI) using Thermo Scientific™ Columbia CNA Aesculin Selective Agar / Brilliance™UTI, a single bi-plate which includes two separate formulations.
Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms while reducing storage capacity issues in the incubator with a bi-plate that includes two widely used microbiological media.
On one side of the plate, Columbia CNA Agar with Asculin isolates Gram-positive organisms from clinical samples with this selective microbiological medium traditionally used to identify staphylococci and streptococci because it provides clear haemolysis and typical growth for S. pneumoniae (dent morphology).
On the other side of the plate, Brilliance UTI isolates, enumerates and presumptively identifies relevant pathogens causing urinary tract infections in 16 to 24 hours. It is a chromogenic medium which differentiates enterococci and Klebsiella spp. easily with distinct color targets which grow on an opaque background. The medium differentiates between coliforms and enterococci, and an improved TDA reaction aids the identification of Proteus, Morganella and Providencia species. Presumptive identification of E.coli can be confirmed using a rapid indole test (DMAC) (MB1448A) for same day results. Chromogens in the formulation help identify key urinary pathogens through the production of distinctive diagnostic colors.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
On one side of the plate, Columbia CNA Agar with Asculin isolates Gram-positive organisms from clinical samples with this selective microbiological medium traditionally used to identify staphylococci and streptococci because it provides clear haemolysis and typical growth for S. pneumoniae (dent morphology).
On the other side of the plate, Brilliance UTI isolates, enumerates and presumptively identifies relevant pathogens causing urinary tract infections in 16 to 24 hours. It is a chromogenic medium which differentiates enterococci and Klebsiella spp. easily with distinct color targets which grow on an opaque background. The medium differentiates between coliforms and enterococci, and an improved TDA reaction aids the identification of Proteus, Morganella and Providencia species. Presumptive identification of E.coli can be confirmed using a rapid indole test (DMAC) (MB1448A) for same day results. Chromogens in the formulation help identify key urinary pathogens through the production of distinctive diagnostic colors.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | CNA: Flame red, opaque. UTI: Ivory, opaque |
| Content And Storage | 6°C to 12°C |
| Description | Columbia CNA Aesculin Selective Agar/Brilliance UTI |
| Format | 90mm biplate |
| Manufacturing Origin | Germany |
| Product Type | Agar |
| Fill Weight | 17g ± 5% |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |