Learn More
Shamrock™ Color Coded Round Adhesive Labels
These round adhesive labels offer universal marking ability with ballpoint pens, pencils and felt pens for easy identification.
14.95 EUR - 16.45 EUR
Specifications
Autoclavable | Autoclavable |
---|---|
For Use With (Application) | Easy identification, labeling |
Labels per Sheet | 1000 |
Shape | Round |
Temperature Range (Metric) | -20°C to +121°C |
Product Code | Brand | Color | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (Metric) | Price | Quantity & Availability | ||||
11786945
|
Shamrock™
SDL-75-2 |
Yellow | 19 mm |
16.45 EUR
Pack of 1000 |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11726955
|
Shamrock™
SDL-75-6 |
Blue | 19 mm |
14.95 EUR
Pack of 1000 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11736955
|
Shamrock™
SDL-75-7 |
Pink | 19 mm |
14.95 EUR
Pack of 1000 |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11796965
|
Shamrock™
SDL-100-12 |
Violet | 25 mm |
15.95 EUR
Pack of 1000 |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11726965
|
Shamrock™
SDL-100-6 |
Blue | 25 mm |
15.95 EUR
Pack of 1000 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11786965
|
Shamrock™
SDL-100-7 |
Pink | 25 mm |
15.95 EUR
Pack of 1000 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Resistant to water, oil and acids
- Temperature range from −20° to +121°C
- Can be used in autoclaves, freezers, refrigerators and ovens
- Provided 1000 labels per sheet
Specifications
Autoclavable | |
1000 | |
-20°C to +121°C |
Easy identification, labeling | |
Round |