Learn More
Collagen IV Antibody (COL4/9395R), Novus Biologicals™
Rabbit Monoclonal Antibody
268.00 EUR - 496.00 EUR
Specifications
| Antigen | Collagen IV |
|---|---|
| Clone | COL4/9395R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625584
|
Novus Biologicals
NBP3-23831-20ug |
20 μg |
268.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639324
|
Novus Biologicals
NBP3-23831-100ug |
100 μg |
496.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Collagen IV Monoclonal antibody specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Collagen IV | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 1282 | |
| IgG κ | |
| Protein A purified |
| COL4/9395R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BSVD;BSVD1;COL4A1s;Collagen alpha-1(IV) chain;PADMAL;RATOR | |
| Recombinant fragment (around aa1400-1670) of human Collagen IV protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |