missing translation for 'onlineSavingsMsg'
Learn More

Colistin, sulfate salt, MP Biomedicals™

Product Code. p-4405695 Shop All MP Biomedicals Products
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11470712

Brand: MP Biomedicals 0219415783

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1264-72-8

Specifications

Color Brown
Quantity 0.25 g
Physical Form Powder
Chemical Name or Material Colistin, sulfate salt
compliance-icons
Product Identifier
  • Colistin Sulfate Salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Colistin Sulfate Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.