Learn More
Invitrogen™ COL9A1 Polyclonal Antibody

Description
Immunogen sequence: WASAAVKRRP RFPVNSNSNG GNELCPKIRI GQDDLPGFDL ISQFQVDKAA SRRAIQRVVG SATLQVAYKL GNNVDFRIPT RNLYPSGLPE EYSFLTTFRM TGSTLKKNWN IWQIQDSSGK EQVGIKINGQ TQSVVFSYKG LDGSLQTAAF SNLSSLFDSQ WHKIMIGVER SSATLFVDCN RIESLPIKPR GPIDIDGFAV LGKLADNPQV SVPFELQWML IHCDPLRPRR ETCHELPARI TPSQTTDERG P; Positive Samples: Mouse skeletal muscle; Cellular Location: extracellular matrix, extracellular space.
Specifications
Specifications
| Antigen | COL9A1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.09 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | COL9A1 |
| Gene Accession No. | P20850 |
| Gene Alias | alpha 1 type IX collagen; alpha-1(IX) collagen chain; cartilage-specific short collagen; COL9A1; Col9a-1; Collagen alpha-1(IX) chain; collagen IX, alpha-1 polypeptide; collagen type IX alpha 1; collagen, type IX, alpha 1; DJ149L1.1.2; EDM6; MED; procollagen, type IX, alpha 1; STL4 |
| Gene Symbols | COL9A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.