Learn More
Sampling Systems™ Cohesive Sampler
Shop All Sampling Systems Products
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
5 mL
10 mL
25 mL
missing translation for 'unitSize'
Each
Description
- Manufactured in 316L Stainless Steel
- Single point type
- Used to push the powder into the sampler by rotating it
Specifications
Specifications
| Product Type | Sampler |
| Material | Stainless Steel (316L) |
| Capacity (Metric) | 10 mL |
| Length (Metric) | 1000 mm |