missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme Q2, TRC

Product Code. 30358658
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358658 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358658 Supplier Toronto Research Chemicals Supplier No. TRCC636450100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Coenzyme Q2
CAS 606-06-4
Molecular Formula C19 H26 O4
Quantity 100 mg
Synonym (E)-2-(3,7-Dimethyl-2,6-octadienyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione, 2-[(2E)-3,7-Dimethyl-2,6-octadienyl]-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione, (E)-2-(3,7-Dimethyl-2,6-octadienyl)-5,6-dimethoxy-3-methyl-p-benzoquinone, 2-[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione, 2-Geranyl-5,6-dimethoxy-3-methyl-p-benzoquinone, 2,3-Dimethoxy-5-geranyl-6-methyl-1,4-benzoquinone, Q 2, Ubiquinone 10, Ubiquinone 2, Ubiquinone Q2
InChI Formula InChI=1S/C19H26O4/c1-12(2)8-7-9-13(3)10-11-15-14(4)16(20)18(22-5)19(23-6)17(15)21/h8,10H,7,9,11H2,1-6H3/b13-10+
SMILES COC1=C(OC)C(=O)C(=C(C)C1=O)C\C=C(/C)\CCC=C(C)C
IUPAC Name 2-[(2E)-3,7-dimethylocta-2,6-dienyl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 318.407
Formula Weight 318.183
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.