missing translation for 'onlineSavingsMsg'
Learn More

Coelenterazine 400a, TRC

Product Code. 30350206
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350206 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350206 Supplier Toronto Research Chemicals Supplier No. TRCC63600325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Coelenterazine 400a
CAS 70217-82-2
Molecular Formula C26 H21 N3 O
Quantity 25 mg
Synonym 6-Phenyl-2,8-bis(phenylmethyl)imidazo[1,2-a]pyrazin-3(7H)-one, Bisdeoxycoelenterazine, Coelenterazine HH, DeepBlueC, Dideoxycoelenterazine
InChI Formula InChI=1S/C26H21N3O/c30-26-23(17-20-12-6-2-7-13-20)28-25-22(16-19-10-4-1-5-11-19)27-24(18-29(25)26)21-14-8-3-9-15-21/h1-15,18,27H,16-17H2
SMILES O=C1N2C=C(NC(=C2N=C1Cc3ccccc3)Cc4ccccc4)c5ccccc5
IUPAC Name 2,8-dibenzyl-6-phenyl-7H-imidazo[1,2-a]pyrazin-3-one
Molecular Weight (g/mol) 391.464
Formula Weight 391.168
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.