Learn More
KARTELL COBAS MIRA™ Multi-Curvette
Click to view available options
Quantity:
1 Segment
15 Segments
Unit Size:
Pack of 450
Description
- 12-position polymethyl methacrylate (PMMA) multiple cuvette segments for the Cobas Mira™; analyzer.
- A high-tech production process and stringent quality control guarantee high reproducibility.
- Available in racks of 15 cuvette segments for devices with continuous throughput or in single segments.
Specifications
Specifications
| Type | Multi-cuvette |
| Sample Type | Whole Blood, Plasma, Serum, Urine |
| Quantity | 15 Segments |