missing translation for 'onlineSavingsMsg'
Learn More

CNBP Antibody (38F10), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-42632PECY55

Product Code. 18199638

  • 547.00 EUR / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CNBP Monoclonal specifically detects CNBP in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CNBP
Monoclonal
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
CNBP
Recombinant human CNBP (1-170aa) purified from E. coli
0.1 mL
Primary
Human
IgG2b κ
Flow Cytometry
38F10
Flow Cytometry
CCHC-type zinc finger, nucleic acid binding protein, cellular nucleic acid binding protein, cellular nucleic acid-binding protein, DM2, erythroid differentiation-related, FLJ11631, PROMM, RNF163CNBP1ZNF9ZCCHC22, sterol regulatory element-binding protein, zinc finger protein 273, Zinc finger protein 9, zinc finger protein 9 (a cellular retroviral nucleic acid binding protein)
Mouse
Protein A purified
DNA replication Transcription Translation and Splicing
7555.0
Store at 4°C in the dark. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.