Learn More
IBA Lifesciences CMV pp50 peptide VTEHDTLLY (HLA-A*0101)
GREENER_CHOICE
Product Code. 15302156 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
15302156 1 mg 1mg
1 options
Product Code. 15302156
Brand: IBA Lifesciences 67024901
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Single peptide synthesized as presented by the MHC class I allele
  • CMV pp50 peptide VTEHDTLLY (HLA-A*0101) is a single peptide for stimulation of T cells.
  • The peptide from human cytomegalovirus (CMV) phosphoprotein (pp50) is synthesized as it is presented by the MHC class I HLA-A*0101 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
  • The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
  • Specifications
    Content And Storage Recommended: -25°C to -15°C
    Quantity 1 mg