Learn More
Clorsulon, MedChemExpress
Product Code. 16468336
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16468336
Brand: Medchem Express HYB0488100mg
MedChemExpress Clorsulon (L631529; MK401) is an orally active flukicidal agent against liver flukes (Fasciola hepatica and Fasciola gigantica) infections in calves and sheep. Clorsulon is also a competitive inhibitor of both 3-phosphoglycorate and ATP andinhibits glucose utilization and acetate and propionate formation by mature Fasciola hepatica in vitro.
Clorsulon (L631529
Specifications
Chemical Name or Material Clorsulon
CAS 60200-06-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C8H8Cl3N3O4S2
Quantity 100 mg
Synonym L631529 MK401
Solubility Information DMSO : 100 mg/mL (262.70 mM; Need ultrasonic)
SMILES O=S(C1=C(/C(Cl)=C(Cl)\Cl)C=C(N)C(S(=O)(N)=O)=C1)(N)=O
Molecular Weight (g/mol) 380.66
Formula Weight 380.66
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation