missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clorgyline Hydrochloride, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Clorgyline Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Clorgyline Hydrochloride |
| CAS | 17780-75-5 |
| Analyte or Component Names | Clorgyline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H15 Cl2 N O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Propyn-1-amine, N-[3-(2,4-dichlorophenoxy)propyl]-N-methyl-, hydrochloride (1:1), 2-Propyn-1-amine, N-[3-(2,4-dichlorophenoxy)propyl]-N-methyl-, hydrochloride (9CI), 2-Propynylamine, N-[3-(2,4-dichlorophenoxy)propyl]-N-methyl-, hydrochloride (7CI,8CI), Clorgyline hydrochloride, N-2,4-Dichlorophenoxypropyl-N-methylpropargylamine-HCl, N-methyl-N-propargyl-3-(2,4-dichlorophenoxy)propylamine hydrochloride |
| InChI Formula | InChI=1S/C13H15Cl2NO.ClH/c1-3-7-16(2)8-4-9-17-13-6-5-11(14)10-12(13)15;/h1,5-6,10H,4,7-9H2,2H3;1H |
| SMILES | Cl.CN(CCCOc1ccc(Cl)cc1Cl)CC#C |
| IUPAC Name | 3-(2,4-dichlorophenoxy)-N-methyl-N-prop-2-ynylpropan-1-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction