missing translation for 'onlineSavingsMsg'
Learn More

Clodronic Acid, TRC

Product Code. 30368570
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30368570 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368570 Supplier Toronto Research Chemicals Supplier No. TRCC58353010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Clodronic Acid
CAS 10596-23-3
Molecular Formula C H4 Cl2 O6 P2
Quantity 10 mg
Synonym Phosphonic acid, P,P'-(dichloromethylene)bis-, Phosphonic acid, (dichloromethylene)bis- (9CI), Phosphonic acid, (dichloromethylene)di- (8CI), P,P'-(Dichloromethylene)bis[phosphonic acid], (Dichloromethylene)bis[phosphonic acid], Cl 2MDP, Clodrolip, Clodro
InChI Formula InChI=1S/CH4Cl2O6P2/c2-1(3,10(4,5)6)11(7,8)9/h(H2,4,5,6)(H2,7,8,9)
SMILES OP(=O)(O)C(Cl)(Cl)P(=O)(O)O
IUPAC Name [dichloro(phosphono)methyl]phosphonic acid
Molecular Weight (g/mol) 244.892
Formula Weight 243.886
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.