missing translation for 'onlineSavingsMsg'
Learn More

Clodronic Acid Disodium Salt Hydrate, TRC

Product Code. 30136231
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136231 10 g 10g
30134204 50 g 50g
17942585 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136231 Supplier Toronto Research Chemicals Supplier No. TRCC58687510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22560-50-5
Molecular Formula C H2 Cl2 O6 P2 . 2 Na
Molecular Weight (g/mol) 288.86
Synonym Phosphonic acid, P,P'-(dichloromethylene)bis-, sodium salt (1:2), Phosphonic acid, (dichloromethylene)bis-, disodium salt (9CI), Phosphonic acid, (dichloromethylene)di-, disodium salt (8CI), (Dichloromethylene)diphosphonate disodium, BM 06011, Bonefos, Clasteon, Clodronate disodium, Clodronate disodium salt, Clodronate sodium, Clodronic acid disodium salt, DClMDP, Dichloromethylenebisphosphonic acid disodium salt, Dichloromethylenediphosphonic acid disodium salt, Difoafonal, Diphosfonal, Disodium (dichloromethane)diphosphonate, Disodium (dichloromethylene)bisphosphonate, Disodium (dichloromethylene)diphosphonate, Disodium clodronate, Lodronate, Loron, Mebonat, Ossiten, Ostac, Sodium (dichloromethylene)diphosphonate, Sodium clodronate
IUPAC Name disodium;[dichloro-[hydroxy(oxido)phosphoryl]methyl]-hydroxyphosphinate
SMILES [Na+].[Na+].OP(=O)([O-])C(Cl)(Cl)P(=O)(O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/CH4Cl2O6P2.2Na/c2-1(3,10(4,5)6)11(7,8)9;;/h(H2,4,5,6)(H2,7,8,9);;/q;2*+1/p-2
Formula Weight 287.8499
Chemical Name or Material Clodronate Disodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.