Learn More
Clinical Standard Sodium 160 mmol/L and Potassium 80 mmol/L for Flame Photometry, Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- Flame Photometry Reference Standard, Flame Photometry Reference Material
- ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
- ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
- ISO/IEC 17025 for temperature calibration in the range of −196 to +1200°C (INAB Ref: 265C)
- ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
- ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
- GHS/OSHA compliant, REACH etc.
- This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of spectroscopy standards), and pre-cleaned bottles.
- The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards where available.
- All of the uncertainties of measurement for this product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.
Specifications
Specifications
| Description | Matrix: Water |
| Packaging | HDPE Triple-leached |
| Analyte or Component Names | Multi-analyte, Sodium, Potassium |
| Physical Form | Liquid |
| Concentration | Mixed concentrations |
| Technique | Flame Photometry |
| Element | Multi-analyte, Na, K |
| Quantity | 500 mL |