Learn More
Nickel Electro™ Clifton™ 56 Litre Stainless Steel Flat Two Piece Lid with Insulated Handles
Click to view available options
For Use With (Equipment):
56 L Clfiton Unstirred Baths
Unit Size:
Each
Description
- Two-part stainless steel flat lids minimize heat loss, save energy, and assist to maintain bath temperature
- Fitted lid acts as a dust cover when Clifton water bath is not in use
- Prevents contamination of liquid from airbourne particles
- The lid is fitted with an insulated handle
Specifications
Specifications
| Description | Stainless Steel flat 2 Pc. lids with insulated handles |
| For Use With (Equipment) | 56 L Clfiton Unstirred Baths |
| Material | Stainless Steel |
| Shipping Weight (Metric) | 2 kg |