Learn More
Corning™ Clear Polystyrene 96-Well EIA/RIA Microplates
Shop All Corning Products
Click to view available options
Well Shape:
Flat Bottom (F-well)
Round
Binding Property:
High Binding
Medium Binding
Packaging:
25/Pk.
Individually Wrapped
Unit Size:
Pack of 100
Pack of 50
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Specifications
| Binding Property | Medium Binding |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Packaging | 25/Pk. |
| Binding Type | Not Treated |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 75 to 200 μL |
| Show More |