Learn More
Hirschmann™ Clear Glass Graduated Pipettes with Amber Stain Graduation
Product Code. 11746632
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
25 mL
50 mL
Graduations:
0.01 mL
0.02 mL
0.05 mL
0.1 mL
0.2 mL
missing translation for 'unitSize'
Pack of 12
Pack of 6
Product Code. 11746632
missing translation for 'mfr': Hirschmann™ 1000142
Only null left
Add to basket
Add to basket
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Clear Glass Graduated Pipettes with Amber Stain Graduation come with a cotton plug end as standard.
  • Short line
  • Acc. to DIN EN ISO 835
  • Graduated to tip, zero on top, type 3
Specifications
Class B
Material Soda Lime Glass
Graduated Yes
Graduation Color Amber
Graduations 0.1 mL
Graduation Type Amber graduations permanently with silver diffusion color
Quantity 12 Pack
Volume (Metric) 2 mL
Length (Metric) 360 ±5 mm
Type Graduated Pipet