Learn More
Cole-Parmer Cleanroom Male Luer Locking Plugs, Straight Adapter
Product Code. 30054044 Shop All Cole Parmer Products
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nylon
Polypropylene
Color:
Clear
Natural
White
Unit Size:
Pack of 10
Product Code. 30054044
Brand: Cole-Parmer 5011049
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Seal luer ports with a secure and leak-proof connection. Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS;USP
Material CrystalVu (CVU)
Inlet Male Luer Lock, Plug
Color Clear
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.