Learn More
Cole-Parmer Cleanroom Large-Bore Female Luer to Hose Barb Fittings, Straight Adapter, Animal-Derivative-Free Polypropylene
Product Code. 30053685 Shop All Cole Parmer Products
Click to view available options
Diameter (English) Outer:
1/4 in.
3/16 in.
5/16 in.
Unit Size:
Pack of 10
Product Code. 30053685
Brand: Cole-Parmer 5010940
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Large-bore luer fittings offer higher flow rates compared to standard luers. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
Material Polypropylene
Diameter (English) Outer 5/16 in.
Packaging Quantity 10/Pk.
Inlet Large Bore Female Luer, Hose Barb