Learn More
Cole-Parmer Cleanroom 10-32 UNF to Hose Barb Fitting, Straight Adapter, Natural Kynar™
Shop All Cole Parmer Products
Click to view available options
Diameter (Metric) Inner:
1/16 in.
1/8 in.
3/32 in.
Thread Size:
10-32
Unit Size:
Pack of 10
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Specifications
Specifications
| Certifications/Compliance | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Diameter (Metric) Inner | 1/16 in. |
| Material | Nylon |
| Packaging Quantity | 10/Pk. |
| Outlet Port | Threaded UNF(M) x Hose Barb |
| Sterilization Method | E-beam/gamma irradiation, EtO, or autoclave up to 121°C |
| Thread Size | 10-32 |
| Shape | Straight |
| Color | White |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.