Learn More
Hirschmann™ Cleaning Detergent rea-clean™
Product Code. 10479322
Click to view available options
Volume (Metric):
1 L
30 L
Unit Size:
Each
Product Code. 10479322
Brand: Hirschmann™ 9790130
Bio degradable cleaning detergent for rapid, automatic and careful cleaning of materials and surfaces especially made of plastics.
  • Biodegradable cleaner
  • Surface cleaning
  • Phosphate-free and self-neutralising
Specifications
Product Type Phosphate-free Cleaning Concentrate
Form Liquid
Product Line rea-clean™
Description Biodegradable, fat dissolving, gentle on materials
For Use With (Equipment) All sufaces in labs
Quantity 1 Can
Volume (Metric) 30 L