Learn More
Sampling Systems™ Cleaning Coupons for Plastics
Shop All Sampling Systems Products
Click to view available options
Material:
HDPE
Nylon
PTFE
PVC
Polypropylene
Dimensions (L x W):
100 x 100 mm
50 x 50 mm
Unit Size:
Each
Description
- Tests available for HDPE, nylon, PP, PTFE and PVC
- Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Specifications
| Product Type | Cleaning Coupon |
| Dimensions (L x W) | 100 x 100 mm |
| For Use With (Application) | Testing and validating cleaning regime |
| Material | PVC |