missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Claudin-6 Antibody (DS-9606a) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
213.00 EUR - 920.00 EUR
Specifications
| Antigen | Claudin-6 |
|---|---|
| Clone | DS-9606a |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044498
|
Novus Biologicals™
NBP3-28463-1mg |
1 mg |
920.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044784
|
Novus Biologicals™
NBP3-28463-100ug |
100 μg |
326.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046007
|
Novus Biologicals™
NBP3-28463-50ug |
50 μg |
213.00 EUR
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Claudin-6 Monoclonal antibody specifically detects Claudin-6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Claudin-6 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Stem Cell Markers | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| claudin 6, claudin-6, skullin | |
| CLDN6 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| DS-9606a | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P56747 | |
| 9074 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title