missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-4 Antibody (10F4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26653-100ul
This item is not returnable.
View return policy
Description
Claudin-4 Monoclonal antibody specifically detects Claudin-4 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| Claudin-4 | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 10F4 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| claudin 4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-RCPETR, CPE-receptor, hCPE-R, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein | |
| A synthesized peptide derived from Human Claudin-4 [UniProt O14493] | |
| 100 μL | |
| Cancer, Extracellular Matrix | |
| 1364 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction