missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26912-1-AP-20UL
This item is not returnable.
View return policy
Description
Claudin 2 Polyclonal antibody apecifically detetects Claudin 2 antigen in Human Samples. It is validated for Western Blot, ELISA.Specifications
| Claudin 2 | |
| Polyclonal | |
| Unconjugated | |
| BC014424 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, ELISA | |
| 650 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| CLDN2 | |
| Claudin 2 fusion protein Ag25527 | |
| 20 μL | |
| 9075 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction