missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Classic Rack for Chest Freezers, with Locking Rod, 32 mm Box height

Product Code. 15200474 Shop All TENAK Products
Click to view available options
Holds:
Boxes (12)
Boxes (16)
Boxes (8)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15200474

Brand: Tenak™ TE21354

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Classic Rack is suitable for use with all chest freezer models.

  • Comes with a locking rod to secure the cryoboxes
  • Measure the inside height of you freezer when choosing the right rack
  • Constructed of stainless steel

Specifications

Material Stainless Steel
For Use With (Equipment) Chest Frseezers
Holds Boxes (12)
Includes Locking rod
Dimensions (L x W x H) 140 x 140 x 340 mm
Height (Metric) 32 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.