Learn More
Tenak™ Classic Rack for Chest Freezers, 100 mm Box Height, with Boxes
Classic Rack is suitable for use with all chest freezer models.
89.75 EUR - 120.00 EUR
Specifications
Includes | White cardboard boxes, 9 x 9 cell dividers |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 100 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15200514
|
Tenak™
TE21474B |
Boxes (6) |
120.00 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
15240504
|
Tenak™
TE21468B |
Boxes (3) |
89.75 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
15260504
|
Tenak™
TE21470B |
Boxes (4) |
98.80 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
15280504
|
Tenak™
TE21472B |
Boxes (5) |
110.00 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
Description
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Stainless Steel | |
Chest Frseezers | |
100 mm (Box) |