Learn More
Tenak™ Classic Rack for Chest Freezers, 100 mm Box Height, with Boxes
Classic Rack is suitable for use with all chest freezer models.
Brand:  Tenak™ TE21468B
Additional Details : Weight : 1.40000kg
 View more versions of this product
Product Code. 15240504
  • 89.75 EUR / Each
Estimated Shipment: 09-08-2024
Description
  • Comes with a locking rod to secure the cryoboxes
  • Measure the inside height of you freezer when choosing the right rack
  • Constructed of stainless steel
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
Specifications
White cardboard boxes, 9 x 9 cell dividers
Stainless Steel
140 x 140 x 320 mm
Boxes (3)
Chest Frseezers
100 mm (Box)